69452-M
HMS174 Competent Cells - Novagen
Escherichia coli, rod shaped
About This Item
Recommended Products
Product Name
HMS174 Competent Cells - Novagen, HMS174 strains provide high transformation efficiencies and the recA mutation in a K-12 background. Strain may stabilize certain target genes whose products may cause the loss of the DE3 prophage.
biological source
Escherichia coli
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
cell transformation
transformation efficiency: >5×106 cfu/μg
shipped in
dry ice
storage temp.
−70°C
Related Categories
General description
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Components
•2 × 0.2 ml5 × 0.2 mlHMS174 Competent Cells
•2 × 2 ml4 × 2 mlSOC Medium
•10 µl10 µlTest Plasmid
Warning
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service