Skip to Content
Merck
All Photos(1)

Key Documents

574777

Sigma-Aldrich

Anti-Synapsin I Rabbit pAb

lyophilized, Calbiochem®

Synonym(s):

Rabbit Anti-Synapsin I, Synapsin I Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

does not contain

preservative

species reactivity

human, rat, mouse, bovine

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Syn1(20964)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the synapsin I protein.
Recognizes the synapsin I protein in rat brain.
This Anti-Synapsin I Rabbit pAb is validated for use in ELISA, Immunoblotting Immunocytochemistry Immunoprecipitation for the detection of Synapsin I.

Immunogen

Bovine
purified, brovine brain synapsin I

Application

ELISA (1:2500-1:10,000)

Frozen Sections (see application references)

Immunoblotting (1:200)

Immunocytochemistry (1:500-1:2500)

Immunoprecipitation (1 µg/200 µg lysate)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 5 mM ammonium bicarbonate.

Reconstitution

Reconstitute with 50 µl PBS. Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
Rat brain

Other Notes

Stone, L.M., et al. 1994. J. Neurosci.14, 301.
Mandell, J.W., et al. 1992. J. Neurosci.12, 1736.
Fletcher, T.L., et al. 1991. J. Neurosci.11, 1617.
Moore, R.Y., and Berstein, M. 1989. J. Neurosci.9, 2151.
This antibody is specific for synapsin I and can be used to estimate synaptic density and synaptogenesis. Strong and specific immunolabeling of the synapsin I doublet seen in immunoblotting at a 1:200 dilution. This immunolabeling can be blocked by preadsorption of antibody with synapsin I. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Koga et al.
Molecular pain, 14, 1744806918783478-1744806918783478 (2018-06-30)
Background Chronic pain is a persistent unpleasant sensation that produces pathological synaptic plasticity in the central nervous system. Both human imaging study and animal studies consistently demonstrate that the anterior cingulate cortex is a critical cortical area for nociceptive and
Valentina N Lagomarsino et al.
Neuron, 109(21), 3402-3420 (2021-09-03)
We have generated a controlled and manipulable resource that captures genetic risk for Alzheimer's disease: iPSC lines from 53 individuals coupled with RNA and proteomic profiling of both iPSC-derived neurons and brain tissue of the same individuals. Data collected for
Bao-Yang Hu et al.
Methods in molecular biology (Clifton, N.J.), 636, 123-137 (2010-03-26)
We describe a chemically defined protocol for efficient differentiation of human embryonic stem cells (hESCs) to neural epithelial cells and then to functional spinal motor neurons. This protocol comprises four major steps. Human ESCs are differentiated without morphogens into neuroepithelial
Loredana Poeta et al.
Genes, 12(7) (2021-08-07)
Dysregulation of transcriptional pathways is observed in multiple forms of neurodevelopmental disorders (NDDs), such as intellectual disability (ID), epilepsy and autism spectrum disorder (ASD). We previously demonstrated that the NDD genes encoding lysine-specific demethylase 5C (KDM5C) and its transcriptional regulators
Madhumita Yennawar et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(24), 4814-4828 (2019-04-07)
Pathogenic mutations in cyclin-dependent kinase-like 5 (CDKL5) result in CDKL5 deficiency disorder (CDD), a rare disease marked by early-life seizures, autistic behaviors, and intellectual disability. Although mouse models of CDD exhibit dendritic instability and alterations in synaptic scaffolding proteins, studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service