Skip to Content
Merck
All Photos(1)

Key Documents

559388

Millipore

SB 202190

≥98% (HPLC), solid, p38 MAP kinase inhibitor, Calbiochem®

Synonym(s):

SB 202190, 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)1H-imidazole, FHPI, p38 MAP Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14FN3O
CAS Number:
Molecular Weight:
331.34
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

SB 202190, SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (Ki = 16 nM; IC₅₀ = 350 nM).

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

16 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C20H14FN3O/c21-16-5-1-13(2-6-16)18-19(14-9-11-22-12-10-14)24-20(23-18)15-3-7-17(25)8-4-15/h1-12,23-24H

InChI key

NJNKPVPFGLGHPA-UHFFFAOYSA-N

General description

A potent, reversible, competitive, and cell-permeable inhibitor of p38 MAP kinase. Inhibits p38 phosphorylation of myelin basic protein (MBP) with no effect on the activity of the ERK or JNK MAP kinase subgroups. Also inhibits the kinase activity of p38β (Ki = 16 nM; IC50 = 350 nM) and p38 phosphorylation of activating transcription factor 2 (ATF-2; IC50 = 280 nM). Blocks LPS-induced TNF-α and interleukin biosynthesis. Reported to induce LDL receptor expression in Hep52 cells.
A potent, reversible, competitive, and cell-permeable inhibitor of p38 MAP kinase. Inhibits p38 phosphorylation of myelin basic protein (MBP) with no effect on the activity of the ERK or JNK MAP kinase subgroups. Also inhibits the kinase activity of p38β (Ki = 16 nM; IC50 = 350 nM) and p38 phosphorylation of activating transcription factor 2 (ATF-2; IC50 = 280 nM). Blocks LPS-induced TNF-α and interleukin biosynthesis. Reported to induce LDL receptor expression in Hep52 cells. A 1 mg/ml solution of SB 202190 (Cat. No. 559397) in anhydrous DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p38β
Product competes with ATP.
Reversible: yes
Target IC50: 350 nM against the kinase activity of p38β; 280 nM against p38 phosphorylation of activating transcription factor 2 (ATF-2)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Davies, S.P., et al. 2000. Biochem. J.351, 95.
Ajizian, S.J., et al. 1999. J. Infect. Dis.179, 939.
Singh, R.P., et al. 1999. J. Biol. Chem.274, 19593.
Wang, S.W., et al. 1999. Inflamm. Res.48, 533.
Gallagher, T.F., et al. 1997. Bioorg. Med. Chem. 5, 49.
LoGrasso, P.V., et al. 1997. Biochem. 36, 10422.
Jiang, Y., et al. 1996. J. Biol. Chem. 271, 17920.
Kramer, R.M., et al. 1996. J. Biol. Chem.271, 27723.
Li, Z., et al. 1996. Biochem. Biophys. Res.Commun. 228, 334.
Lee, J.C., et al. 1994. Nature 372, 739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brooke Wang et al.
Current protocols, 4(4), e1027-e1027 (2024-04-08)
The development of patient-derived intestinal organoids represents an invaluable model for simulating the native human intestinal epithelium. These stem cell-rich cultures outperform commonly used cell lines like Caco-2 and HT29-MTX in reflecting the cellular diversity of the native intestinal epithelium
Christina L Sherry et al.
Journal of immunology (Baltimore, Md. : 1950), 178(2), 663-670 (2007-01-05)
Dysregulated inflammation is a complication of type 2 diabetes (T2D). In this study, we show that augmented LPS-induced TNF-alpha production by resident peritoneal macrophages (PerMphi) in type 2 diabetic (db/db) mice is dependent on elevated glucose and requires p38 MAPK.
Trine Bertelsen et al.
Annals of dermatology, 33(2), 122-130 (2021-05-04)
Interleukin (IL)-19 and IL-20 are important members of the IL-10 cytokine family, which are known to play a role in inflammatory processes. Both anti-IL-19 and -IL-20 targeting drugs have been suggested in the treatment of inflammatory diseases such as psoriasis
Muya Zhou et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 10(25), e2301059-e2301059 (2023-07-03)
When circulating tumor cells (CTCs) travel in circulation, they can be killed by detachment-induced anoikis and fluidic shear stress (SS)-mediated apoptosis. Circulatory treatment, which can make CTCs detached but also generate SS, can increase metastasis of cancer cells. To identify
Maike Mose et al.
Experimental dermatology, 22(7), 476-481 (2013-06-27)
The antimicrobial peptide S100A8 is known to be upregulated in lesional psoriatic skin compared with non-lesional psoriatic skin and is believed to play a role in the pathogenesis of psoriasis. However, little is known about the signalling pathways involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service