Skip to Content
Merck
All Photos(1)

Key Documents

553400

Sigma-Aldrich

Radicicol, Diheterospora chlamydosporia

Synonym(s):

Radicicol, Diheterospora chlamydosporia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17ClO6
CAS Number:
Molecular Weight:
364.78
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

description

Merck USA index - 14, 6253

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

0.27 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c2c(c(cc1O)O)C(=O)OC(CC3OC3\C=C\C=C\C(=O)C2)C

InChI

1S/C18H17ClO6/c1-9-6-15-14(25-15)5-3-2-4-10(20)7-11-16(18(23)24-9)12(21)8-13(22)17(11)19/h2-5,8-9,14-15,21-22H,6-7H2,1H3/b4-2+,5-3+

InChI key

WYZWZEOGROVVHK-ZUVMSYQZSA-N

General description

A cell-permeable, ATP-site binding, and irreversible antifungal macrocyclic lactone antibiotic that acts as a protein tyrosine kinase inhibitor. Inhibits p60v-src kinase activity (IC50 = 0.27 µM). Inhibits the expression of COX-2 (IC50 = 27 nM) without affecting COX-1 expression in LPS-stimulated macrophages. Disrupts k-Ras-activated signaling pathway by selectively depleting Raf kinase. Inhibits tyrosine phosphorylation of p53/56lyn in LPS-stimulated macrophages. Suppresses NIH 3T3 cell transformation by diverse oncogenes, such as src, ras and mos, at least in part by blocking the key signal transduction intermediates, such as MAP kinase and GAP-associated p62. Also exhibits anti-angiogenic activity in vivo. Inhibits Wnt-5A expression in dermal papilla cells (IC50 = 190 nM).

Biochem/physiol Actions

Primary Target
p60v-src kinase activity
Secondary Target
COX-2 (IC₅₀ = 27 nM)

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Other Notes

Shinonaga, H., et al. 2009. Bioorg. Med. Chem.17, In press.
Soga, S., et al. 1998. J. Biol. Chem. 273, 822.
Chanmugam, P., et al. 1995. J. Biol. Chem. 270, 5418.
Shimada, Y., et al. 1995. J. Antibiot. 48, 824.
Zhao, J.F., et al. 1995. Oncogene 11, 161.
Oikawa, T., et al. 1993. Eur. J. Pharmacol. 241, 221.
Kwon, H.J., et al. 1992. Cancer Res. 52, 6926.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingshe Han et al.
Frontiers in genetics, 11, 541944-541944 (2020-12-22)
Accumulating evidence indicates that heat shock protein 90 (HSP90) plays essential roles in modulation of phenotypic plasticity in vertebrate development, however, the roles of HSP90 in modulation of cold tolerance capacity in fish are still unclear. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service