Skip to Content
Merck
All Photos(1)

Key Documents

539522

Sigma-Aldrich

PKCε Translocation Inhibitor Peptide

The PKCε Translocation Inhibitor Peptide, Negative Control controls the biological activity of PKCε. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKCε Translocation Inhibitor Peptide, Protein Kinase Cε Translocation Inhibitor I, EAVSLKPT, PKCε Translocation Inhibitor I, PKCε-RACK2 Interaction Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H65N9O13
Molecular Weight:
843.96
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

An octapeptide that selectively inhibits the translocation of protein kinase Cε to subcellular sites in a reversible manner. Inhibition of PKCε translocation is known to specifically block phorbol ester- or norepinephrine-mediated regulation of contraction in cardiomyocytes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PKCε Translocation
Product does not compete with ATP.
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Glu-Ala-Val-Ser-Leu-Lys-Pro-Thr-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yedovitzky, M., et al. 1997. J. Biol. Chem. 272, 1417.
Johnson, J.A., et al. 1996. J. Biol. Chem.271, 24962.

Legal Information

Sold under license of U.S. Patent 5,783,405.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wattamon Srisakuldee et al.
Cells, 10(10) (2021-10-24)
Mitochondria, abundant organelles in high energy demand cells such as cardiomyocytes, can determine cell death or survival by regulating the opening of mitochondrial permeability transition pore, mPTP. We addressed the hypothesis that the growth factor FGF2, known to reside in
Neus Garcia et al.
Molecular neurobiology, 59(7), 4044-4064 (2022-04-28)
During the nervous system development, synapses are initially overproduced. In the neuromuscular junction (NMJ) however, competition between several motor nerve terminals and the synapses they made ends with the maturation of only one axon. The competitive signaling between axons is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service