Skip to Content
Merck
All Photos(1)

Key Documents

5.08017

Sigma-Aldrich

nor-Binaltorphimine Dihydrochloride

≥98% (HPLC), solid, κ-opioid receptor antagonist, Calbiochem

Synonym(s):

nor-Binaltorphimine Dihydrochloride, Norbinaltorphimine, nor-BNI, κ Opioid Receptor Antagonist, nor-Binaltorphimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H43N3O6 · 2HCl
CAS Number:
Molecular Weight:
734.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

nor-Binaltorphimine Dihydrochloride,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

water: 50 mM

storage temp.

−20°C

SMILES string

N21CC3(C4(C(Oc%12c4c(ccc%12O)CC2C%13CC%13)c5[nH]c6c(c5C3)CC7(C98C6Oc%10c9c(ccc%10O)CC(N(C7)CC8)C%11CC%11)O)CC1)O

InChI

1S/C40H43N3O6/c44-27-7-5-21-13-25(19-1-2-19)42-11-9-39-29(21)33(27)48-35(39)31-23(15-37(39,46)17-42)24-16-38(47)18-43-12-10-40(38)30-22(14-26(43)20-3-4-20)6-8-28(45)34(30)49-36(40)32(24)41-31/h5-8,19-20,25-26,35-36,41,44-47H,1-4,9-18H2

InChI key

GYEXSFKLGQBRBJ-UHFFFAOYSA-N

General description

A potent antagonist highly selective for κ opioid receptor with no effect on µ- or &delta- opioid receptors. Widely used in studies related to pain, addition, anxiety, depression, and sleep.

Biochem/physiol Actions

Primary Target
kappa opioid receptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kishioka, S., et al. 2013. Neurosci. Letters.In press.

Valdez, G., et al. 2012. Pharma. Biochem. and Behavior.102, 44.
Takemori AE et al. 1988. J. Pharmacol. Exp. Ther.246, 255.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service