5.00561
Ezh2 Inhibitor II, EI1
Synonym(s):
Ezh2 Inhibitor II, EI1, Enhancer of Zested Homolog 2 Inhibitor II, HMTase Inhibitor XI, 6-Cyano-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-1-(pentan-3-yl)-1H-indole-4-carboxamide
About This Item
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 10 mg/mL
storage temp.
2-8°C
SMILES string
[nH]1[c](c(c(cc1C)C)CNC(=O)c2c3c([n](cc3)C(CC)CC)cc(c2)C#N)=O
InChI
1S/C23H26N4O2/c1-5-17(6-2)27-8-7-18-19(10-16(12-24)11-21(18)27)22(28)25-13-20-14(3)9-15(4)26-23(20)29/h7-11,17H,5-6,13H2,1-4H3,(H,25,28)(H,26,29)
InChI key
PFHDWRIVDDIFRP-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Ezh2/PRC2
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service