Skip to Content
Merck
All Photos(1)

Key Documents

493800

Sigma-Aldrich

PARP Inhibitor VI, NU1025

The PARP Inhibitor VI, NU1025, also referenced under CAS 90417-38-2, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor VI, NU1025, 8-Hydroxy-2-methylquinazoline-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O2
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[nH]1[c](c2c(nc1C)c(ccc2)O)=O

InChI

1S/C9H8N2O2/c1-5-10-8-6(9(13)11-5)3-2-4-7(8)12/h2-4,12H,1H3,(H,10,11,13)

InChI key

YJDAOHJWLUNFLX-UHFFFAOYSA-N

General description

A potent inhibitor poly(ADP-ribose) polymerase (PARP; IC50 = 0.40 µM). Has been shown to potentiate the cytotoxicity of the DNA-methylating agent MTIC [5-(3-N-methyltriazen-1-yl)-imidazole-4-carboxamide] and ionizing irradiation in murine L210 leukemia cells.
A potent poly(ADP-ribose) polymerase (PARP) inhibitor (IC50 = 400 nM) that potentiates the cytotoxicity of various DNA-active agents, including the DNA-methylating compound MTIC [5-(3-N-methyltriazen-1-yl)-imidazole-4-carboxamide], the DNA strand break-inducing drug temozolomide, topotecan, bleomycin, and ionizing radiation in murine L1210 leukemia cells, Chinese hamster ovary cells, and in a variety of human tumor cell lines.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PARP
Product does not compete with ATP.
Reversible: no
Target IC50: 0.4 µM against poly(ADP-ribose) polymerase (PARP)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

This solution can be further diluted 1:100 in tissue culture medium before use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Delaney, C.A., et al. 2000. Clin. Cancer Res.6, 2860.
Boulton, S., et al. 1999. Carcinogenesis20, 199.
Bowman, K.J., et al. 1998. Br. J. Cancer 78, 1269.
Griffin, R.J., et al. 1998. J. Med. Chem. 41, 5247.
Griffin, R.J., et al. 1996. Pharm. Sci. 2, 43.
Boulton, S., et al. 1995. Br. J. Cancer 72, 849.
Griffin, R.J., et al. 1995. Anticancer Drug Res. 10, 507.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service