444283
MMP-13 Inhibitor
The MMP-13 Inhibitor, also referenced under CAS 544678-85-5, controls the biological activity of MMP-13. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Synonym(s):
MMP-13 Inhibitor, Pyrimidine-4,6-dicarboxylic acid, bis-(4-fluoro-3-methyl-benzylamide)
About This Item
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
light yellow
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
Fc1c(cc(cc1)CNC(=O)c2ncnc(c2)C(=O)NCc3cc(c(cc3)F)C)C
InChI
1S/C22H20F2N4O2/c1-13-7-15(3-5-17(13)23)10-25-21(29)19-9-20(28-12-27-19)22(30)26-11-16-4-6-18(24)14(2)8-16/h3-9,12H,10-11H2,1-2H3,(H,25,29)(H,26,30)
InChI key
PYFRREJCFXFNRR-UHFFFAOYSA-N
General description
Biochem/physiol Actions
MMP-13 activity
Packaging
Warning
Preparation Note
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service