Skip to Content
Merck
All Photos(1)

Key Documents

420305

Sigma-Aldrich

K-252c

A cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (IC₅₀ = 2.45 µM) and protein kinase A (IC₅₀ = 25.7 µM).

Synonym(s):

K-252c

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N3O
CAS Number:
Molecular Weight:
311.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 5 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2c3[nH]c4c(c3c5c(c2c6c1cccc6)CNC5=O)cccc4

InChI

1S/C20H13N3O/c24-20-17-12(9-21-20)15-10-5-1-3-7-13(10)22-18(15)19-16(17)11-6-2-4-8-14(11)23-19/h1-8,22-23H,9H2,(H,21,24)

InChI key

MEXUTNIFSHFQRG-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase A (IC50 = 25.7 µM) and protein kinase C (IC50 = 2.45 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product competes with ATP.
Reversible: yes
Target IC50: 2.45 µM against protein kinase C

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Fabre, S., et al. 1993. Bioorg. Med. Chem.1, 193.
Nakanishi, S., et al. 1986. J. Antibiot. 39, 1066.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service