Skip to Content
Merck
All Photos(1)

Key Documents

405268

Sigma-Aldrich

Indomethacin

≥98% (HPLC), powder, COX inhibitor, Calbiochem

Synonym(s):

Indomethacin, 1-( p-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic Acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Indomethacin, A non-steroidal anti-inflammatory, cell permeable, antipyretic agent.

Quality Level

description

Merck USA index - 14, 4968

Assay

≥98% (by assay)

form

powder

potency

740 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

ethanol: 20 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

Clc1ccc(cc1)C(=O)[n]2c3c(c(c2C)CC(=O)O)cc(cc3)OC

InChI

1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)

InChI key

CGIGDMFJXJATDK-UHFFFAOYSA-N

General description

A cell-permeable, non-steroidal anti-inflammatory, anti-pyretic agent. Non-selective cyclooxygenase (COX) inhibitor (IC50 = 740 nM for COX-1; IC50 = 970 nM for COX-2). Inhibits phospholipase A2 (IC50 = 145 µM). Reported to reduce Aβ42 levels independently of COX activity.
A non-steroidal anti-inflammatory, cell permeable, antipyretic agent. Non-selective COX inhibitor (IC50 = 740 nM for COX-1; IC50 = 970 nM for COX-2). Inhibits phospholipase A2 (IC50 = 145 µM). Strongly inhibits insoluble transthyretin (TTR) amyloid fibril formation and suppresses production of Aβ-peptide and secreted form of APP by inhibition of APP mRNA levels in NG108-15 cells.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kodoyama, K., et al. 2001. Biochem. Biophys. Res. Commun.281, 483.
Weggen, S., et al. 2001. Nature414, 212.
Kalgutkar, A.S., et al. 2000. Proc. Natl. Acad. Sci. USA97, 925.
Klabunde, T., et al. 2000. Nat. Struct. Biol.7, 312.
Futaki, N., et al. 1994. Prostaglandins47, 55.
Futaki, N., et al. 1994. Prostaglandins47, 55.
Stevenson, K.M., and Lumbers, E.R. 1992. J. Dev. Physiol. 17, 257.
Oliw, E. 1980. Prostaglandins19, 271.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kadoyama et al.
Biochemical and biophysical research communications, 281(2), 483-490 (2001-02-22)
Cyclooxygenase (COX) synthesizes bioactive prostaglandins from arachidonic acid, and there are COX-1 and COX-2 isoforms with distinct pathophysiological functions. Recent studies demonstrated that COX-2 expression was up-regulated in the brain of patients with Alzheimer's disease. We established mouse neuroblastoma x
K M Stevenson et al.
Journal of developmental physiology, 17(6), 257-264 (1992-06-01)
The effects of indomethacin (10 mg/kg i.v. to the ewe and 12 mg/kg i.v. to the fetus) were examined in 8 chronically catheterized fetal sheep (117-138 days gestation). These doses suppressed fetal 6-keto-prostaglandin F1 alpha and thromboxane B2 levels. Fetal
Sai Koung Ngeun et al.
Biology, 13(2) (2024-02-23)
The myogenic differentiation potential of MSCs is a key factor in their potential use as a cell source for muscle tissue repair and regeneration. Additionally, evaluating the immunomodulatory properties of MSCs is important to highlight their potential for regulating inflammation
S Weggen et al.
Nature, 414(6860), 212-216 (2001-11-09)
Epidemiological studies have documented a reduced prevalence of Alzheimer's disease among users of nonsteroidal anti-inflammatory drugs (NSAIDs). It has been proposed that NSAIDs exert their beneficial effects in part by reducing neurotoxic inflammatory responses in the brain, although this mechanism
Ahlam M Abdallah et al.
Naunyn-Schmiedeberg's archives of pharmacology (2024-07-29)
The type II collagen-induced arthritis (CIA) model and human rheumatoid arthritis exhibit similar characteristics. Both diseases involve the production of inflammatory cytokines and other mediators, triggering an inflammatory cascade linked to bone and cartilage damage. Recently, new pyrazole compounds with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service