Skip to Content
Merck
All Photos(1)

Key Documents

364205

Sigma-Aldrich

GM 6001

≥95% (HPLC), solid, Matrix metalloproteinase inhibitor, Calbiochem

Synonym(s):

GM 6001, Galardin, N-[(2R)-2-(Hydroxamidocarbonylmethyl)-4-methylpentanoyl]-L-tryptophan Methylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N4O4
CAS Number:
Molecular Weight:
388.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

GM 6001, GM 6001, CAS 142880-36-2, is a potent, cell-permeable, broad-spectrum inhibitor of matrix metalloproteinases.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[nH]1c2c(c(c1)C[C@H](NC(=O)[C@H](CC(C)C)CC(=O)NO)C(=O)NC)cccc2

InChI

1S/C20H28N4O4/c1-12(2)8-13(10-18(25)24-28)19(26)23-17(20(27)21-3)9-14-11-22-16-7-5-4-6-15(14)16/h4-7,11-13,17,22,28H,8-10H2,1-3H3,(H,21,27)(H,23,26)(H,24,25)/t13-,17+/m1/s1

InChI key

NITYDPDXAAFEIT-DYVFJYSZSA-N

General description

A cell-permeable, potent broad-spectrum hydroxamic acid inhibitor of matrix metalloproteinases (MMPs). [Ki = 0.4 nM for skin fibroblast collagenase (MMP-1); Ki = 0.5 nM for gelatinase A (MMP-2); Ki = 27 nM for stromelysin (MMP-3); Ki = 0.1 nM for neutrophil collagenase (MMP-8); and Ki = 0.2 nM for gelatinase B (MMP-9)]. Also prevents the release of TNF-α in vivo and in vitro and abrogates endotoxin-induced lethality in mice.
A potent, cell-permeable, broad-spectrum hydroxamic acid inhibitor of matrix metalloproteinases (MMPs); (Ki = 400 pM for MMP-1; Ki = 500 pM for MMP-2; Ki = 27 nM for MMP-3; Ki = 100 pM for MMP-8; and Ki = 200 pM for MMP-9). Prevents the release of TNF-α in vivo and in vitro and abrogates endotoxin-induced lethality in mice. A 10 mM (1 mg/257 µl) solution of GM6001 (Cat. No. 364206) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-1
Product does not compete with ATP.
Reversible: no
Target Ki: 400 pM for MMP-1; 500 pM for MMP-2; 27 nM for MMP-3; 100 pM for MMP-8; 200 pM for MMP-9

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Solorzano, C.C., et al. 1997. Shock7, 427.
Galardy, R.E., et al. 1994. Ann. NY Acad. Sci.732, 315.
Galardy, R.E., et al. 1994. Cancer Res.54, 4715.
Galardy, R.E. 1993. Drugs Future18, 1109.
Grobelny, D., et al. 1992. Biochemistry31, 7152.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Liu et al.
Brain : a journal of neurology, 145(11), 4108-4123 (2022-01-19)
Chronic pain is the most common neurological disorder of HIV patients. Multiple neuropathologies were identified in the pain pathway. Among them is the prominent astrocytic reaction (also know an astrogliosis). However, the pathogenic role and mechanism of the astrogliosis are
Amélie Robert et al.
Methods in molecular biology (Clifton, N.J.), 788, 219-247 (2011-12-02)
The cloning of thrombopoietin together with advances in the culture of hematopoietic stem cells have paved the way for the study of megakaryopoiesis, ongoing clinical trials and, in the future, for the potential therapeutic use of ex vivo produced blood
Hui Geng et al.
American journal of physiology. Renal physiology, 320(3), F359-F374 (2021-01-12)
Lysophosphatidic acid (LPA) increases platelet-derived growth factor-B (PDGFB) and connective tissue growth factor (CTGF) production and secretion by proximal tubule (PT) cells through LPA2 receptor-Gqα-αvβ6-integrin-mediated activation of transforming growth factor-β1 (TGFB1). LPA2, β6-integrin, PDGFB, and CTGF increase in kidneys after
Sandra C Côté et al.
European journal of immunology, 50(10), 1537-1549 (2020-05-12)
The IL-7 receptor specific α chain, CD127, can be expressed both as a membrane-associated (mCD127) and a soluble form (sCD127), however, the mechanisms involved in their regulation remain to be defined. We first demonstrated in primary human CD8+ T cells
Nicolas Pineault et al.
Methods in molecular biology (Clifton, N.J.), 946, 205-224 (2012-11-28)
Megakaryocytes (MK) are hematopoietic cells present in the bone marrow that are responsible for the production and release of platelets in the circulation. Given their very low frequency (<1%), human MK often need to be derived in culture to study

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service