Skip to Content
Merck
All Photos(1)

Key Documents

354400

Sigma-Aldrich

Glutaraldehyde Solution

25% Aqueous

Synonym(s):

Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.32
technique(s):
electron microscopy: suitable

Product Name

Glutaraldehyde, 25% Aqueous Solution,

description

Merck USA index - 14, 4472

Quality Level

Assay

≥98% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

electron microscopy: suitable

color

clear light yellow

solubility

water: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues.

Warning

Toxicity: Toxic (F)

Physical form

25% aqueous solution.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sujin Park et al.
Histochemistry and cell biology, 153(2), 89-99 (2019-11-14)
Autophagy is a degradative cellular process that can be both non-selective and selective and begins with the formation of a unique smooth double-membrane phagophore which wraps around a portion of the cytoplasm. Excess and damaged organelles and cytoplasmic protein aggregates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service