Skip to Content
Merck
All Photos(1)

Key Documents

33-016

Sigma-Aldrich

PI 3-Kinase HTRF® Assay; 384 wells

The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 1 plate.

Synonym(s):

Assay for PI 3-Kinase, HTRF Assay, PI 3-Kinase Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human, mouse

manufacturer/tradename

HTRF®
Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

detection method

fluorometric (HTRF)

shipped in

dry ice

Gene Information

human ... PIK3CA(5290)

Application

Used to detect/quantify: PI 3-Kinase

Biochem/physiol Actions

Target Sub-Family: PI3K/PI4K

Packaging

Kit capacity: 384 wells

Components

4X Reaction Buffer

PIP2 (Substrate; 1mM stock)

Stop Reagent A

Stop Reagent B

DM A Detection Reagent

DM B Detection Reagent

DM C Detection Reagent

Legal Information

HTRF is a registered trademark of Cis Bio International
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James M J Dickson et al.
Bioscience reports, 33(6) (2013-08-24)
NTT (N-terminal tags) on the catalytic (p110) sub-unit of PI 3-K (phosphoinositol 3-kinase) have previously been shown to increase cell signalling and oncogenic transformation. Here we test the impact of an NT (N-terminal) His-tag on in vitro lipid and protein
Christina M Buchanan et al.
PloS one, 8(8), e71337-e71337 (2013-08-13)
In addition to lipid kinase activity, the class-I PI 3-kinases also function as protein kinases targeting regulatory autophosphorylation sites and exogenous substrates. The latter include a recently identified regulatory phosphorylation of the GM-CSF/IL-3 βc receptor contributing to survival of acute
Stephen Jamieson et al.
The Biochemical journal, 438(1), 53-62 (2011-06-15)
Genetic alterations in PI3K (phosphoinositide 3-kinase) signalling are common in cancer and include deletions in PTEN (phosphatase and tensin homologue deleted on chromosome 10), amplifications of PIK3CA and mutations in two distinct regions of the PIK3CA gene. This suggests drugs
Qin Li et al.
Toxicology research, 8(3), 353-360 (2019-06-05)
Qingkailing injection (QKLI) is a kind of multi-component traditional Chinese medicine injection. It has been widely used in clinical practice, but in recent years, it has caused more and more adverse reactions, mainly manifested as pseudo-allergic symptoms. To explore the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service