Skip to Content
Merck
All Photos(1)

Key Documents

324623

Sigma-Aldrich

Eg5 Inhibitor IV, VS-83

The Eg5 Inhibitor IV, VS-83, also referenced under CAS 909250-29-9, controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Eg5 Inhibitor IV, VS-83, 5-Fluoro-3,4-dihydro-4(3ʹ-hydroxyphenyl)quinazoline-2(1H)-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11FN2OS
CAS Number:
Molecular Weight:
274.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 5 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1c2c(ccc1)NC(=S)NC2c3cc(ccc3)O

InChI

1S/C14H11FN2OS/c15-10-5-2-6-11-12(10)13(17-14(19)16-11)8-3-1-4-9(18)7-8/h1-7,13,18H,(H2,16,17,19)

InChI key

MAEYCDNDKNPNQJ-UHFFFAOYSA-N

General description

A cell-permeable Monastrol (Cat. No. 475879) analog with much enhanced potency and selectivity (over 8 other kinesins) in inhibiting mitotic kinesin Eg5-ATPase activity (IC50 = 1.17 vs. 20.73 µM for VS-83 and Monastrol, respectively) and in inducing monoasters formation (EC50 = 7.27 vs. 58.74 µM for BSC-1 cells treated with VS-83 and Monastrol, respectively). Shown to exhibit ≥10-fold antiproliferative activity than Monastrol against U-87 MG, U-118 MG, and U-373 MG glioblastoma cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Muller, C., et al. 2007. Cancer Chemother. Pharmacol.59, 157.
Santamaria, A., et al. 2007. Mol. Biol. Cell18, 4024.
Sarli, V., et al. 2005. ChemBioChem6, 2005.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service