Skip to Content
Merck
All Photos(1)

Key Documents

3030-OP

Millipore

Cesium Chloride

OmniPur® Grade, >= 99.5%

Synonym(s):

OmniPur® Cesium Chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClCs
CAS Number:
Molecular Weight:
168.36
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
form:
crystals

Quality Level

form

crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

SMILES string

[Cs+].[Cl-]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Related Categories

General description

Molecular biology grade.





Specifications:




A (260 nm, 50%, water): ≤0.1AU




Assay: ≥99.5%




DNase: None detected




Lead (Pb): ≤0.001%




Lithium (Li): ≤0.005%




Magnesium (Mg): ≤0.001%




Nickel (Ni): ≤0.0001%




Potassium (K): ≤0.05%




Protease: None detected




RNase: None detected




Rubidium (Rb): ≤0.1%




Sodium (Na): ≤0.1%









Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Nanostructured Polyoxometalate-Based Heterogeneous Electrode Materials for Electrochemical Sensing of Glucose.: The research demonstrates the use of cesium chloride in the development of advanced electrode materials for glucose sensing, enhancing diabetes management technologies (Yu et al., 2024).


Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service