Skip to Content
Merck
All Photos(1)

Key Documents

234140

Sigma-Aldrich

Collagenase Inhibitor I

The Collagenase Inhibitor I controls the biological activity of Collagenase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Collagenase Inhibitor I, Z-PDLDA-NHOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32N4O6
Molecular Weight:
448.51
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

1 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent and specific inhibitor of vertebrate collagenases with IC50 values in the order of 1 µM.

Biochem/physiol Actions

Cell permeable: no
Primary Target
vertebrate collegenases
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Pro-D-Leu-D-Ala-NHOH

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Odake, S., et al. 1990. Chem. Pharm. Bull. 38, 1007.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service