Skip to Content
Merck
All Photos(2)

Key Documents

217694

Sigma-Aldrich

Cdc25 Inhibitor IV, NSC 95397

The Cdc25 Inhibitor IV, NSC 95397, also referenced under CAS 93718-83-3, controls the biological activity of Cdc25. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdc25 Inhibitor IV, NSC 95397, (2,3-bis-[2-hydroxyethylsulfanyl]-[1,4]naphthoquinone), PTP Inhibitor XXIX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O4S2
CAS Number:
Molecular Weight:
310.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 100 mg/mL, clear, red

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C1C(SCCO)=C(SCCO)C(C2=CC=CC=C21)=O

InChI

1S/C14H14O4S2/c15-5-7-19-13-11(17)9-3-1-2-4-10(9)12(18)14(13)20-8-6-16/h1-4,15-16H,5-8H2

InChI key

MAASHDQFQDDECQ-UHFFFAOYSA-N

General description

A cell-permeable, para-naphthoquinone that has been reported to inhibit protein tyrosine phosphatase activity, including Cdc25 dual specificity phosphatase (Ki = 32 nM, 96 nM, and 40 nM for Cdc25A, -B, and -C, respectively) and blocks G2M transition in murine carcinoma cells. In addition, it is shown to inhibit S100A4 binding to myosin-IIA via covalent modification of cysteine residues, Cys81 and Cys86, in the target binding cleft (IC50 = 1.8 µM). It demonstrates inhibitory activity against S100A4-mediated depolymerization of myosin-IIA filaments.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dulyaninova, N.G., et al. 2011. Biochemistry50, 7218.
Lazo, J.S., et al. 2002. Mol. Pharmacol.61, 720.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucia Falbo et al.
iScience, 27(8), 110567-110567 (2024-08-26)
Replication origin assembly is a pivotal step in chromosomal DNA replication. In this process, the ORC complex binds DNA and, together with the CDC6 and CDT1, promotes the loading of the MCM helicase. Chemicals targeting origin assembly might be useful

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service