Skip to Content
Merck
All Photos(1)

Key Documents

21-169

Sigma-Aldrich

FOPflash (mutant TCF binding sites)

Transfection grade T cell factor (TCF) reporter plasmid containing 2 full & one incomplete copy of the Tcf binding site (mutated) followed by 3 copies in the reverse orientation. Serves as a negative control to TOPflash.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106617
eCl@ss:
34360190
NACRES:
NA.41

Quality Level

manufacturer/tradename

Upstate®

technique(s)

transfection: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-18/WNT1-inducible signaling pathway protein-1 signaling mediates human saphenous vein smooth muscle cell proliferation.
Venkatapuram Seenu Reddy,Anthony J Valente,Patrice Delafontaine,Bysani Chandrasekar
Journal of Cellular Physiology null
M van de Wetering et al.
Cell, 88(6), 789-799 (1997-03-21)
The vertebrate transcription factors TCF (T cell factor) and LEF (lymphocyte enhancer binding factor) interact with beta-catenin and are hypothesized to mediate Wingless/Wnt signaling. We have cloned a maternally expressed Drosophila TCF family member, dTCF. dTCF binds a canonical TCF
V Korinek et al.
Molecular and cellular biology, 18(3), 1248-1256 (1998-03-06)
Tcf transcription factors interact with beta-catenin and Armadillo to mediate Wnt/Wingless signaling. We now report the characterization of genes encoding two murine members of the Tcf family, mTcf-3 and mTcf-4. mTcf-3 mRNA is ubiquitously present in embryonic day 6.5 (E6.5)
T Ishitani et al.
Nature, 399(6738), 798-802 (1999-07-03)
The Wnt signalling pathway regulates many developmental processes through a complex of beta-catenin and the T-cell factor/lymphoid enhancer factor (TCF/LEF) family of high-mobility-group transcription factors. Wnt stabilizes cytosolic beta-catenin, which then binds to TCF and activates gene transcription. This signalling
M P Coghlan et al.
Chemistry & biology, 7(10), 793-803 (2000-10-18)
Glycogen synthase kinase-3 (GSK-3) is a serine/threonine protein kinase, the activity of which is inhibited by a variety of extracellular stimuli including insulin, growth factors, cell specification factors and cell adhesion. Consequently, inhibition of GSK-3 activity has been proposed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service