Skip to Content
Merck
All Photos(1)

Key Documents

208719

Sigma-Aldrich

ALLN

≥95% (HPLC), solid, calpain inhibitor, Calbiochem®

Synonym(s):

ALLN, LLNL, MG 101, Calpain Inhibitor I, Proteasome Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O4
CAS Number:
Molecular Weight:
383.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

ALLN, Cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H37N3O4/c1-7-8-9-16(12-24)22-19(26)18(11-14(4)5)23-20(27)17(10-13(2)3)21-15(6)25/h12-14,16-18H,7-11H2,1-6H3,(H,21,25)(H,22,26)(H,23,27)/t16-,17-,18-/m0/s1

InChI key

FMYKJLXRRQTBOR-BZSNNMDCSA-N

General description

Cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM). Inhibits neutral cysteine proteases and the proteasome (Ki = 6 µM). Modulates the processing of the β-amyloid precursor protein (βAPP) to β-amyloid (Aβ). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Also inhibits reovirus-induced apoptosis in L929 cells. Inhibits the proteolysis of IκB-α and IκB-β by the ubiquitin-proteasome complex. Inhibits cell cycle progression at G1/S and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene. A 10 mM (5 mg/1.30 ml) solution of ALLN (Cat. No. 208750) in DMSO is also available.
Inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM). Inhibits neutral cysteine proteases and proteasome (Ki = 6 µM). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Inhibits cell cycle progression at the G1/S border and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene. Inhibits the proteolysis of IκBα and IκBβ by the ubiquitin-proteasome complex.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Calpain-1
Product does not compete with ATP.
Reversible: no
Target Ki: 190 nM, 220 nM, 150 nM, 500 pM, against calpain I, calpain II, cathepsin B, and cathepsin L, respectively

Warning

Toxicity: Standard Handling (A)

Sequence

N-Acetyl-Leu-Leu-Nle-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Debiasi, R.L., et al. 1999. J. Virol. 73, 695.
Zhang, L., et al. 1999. J. Biol. Chem.274, 8966.
Milligan, S.A., et al. 1996. Arch. Biochem. Biophys. 335, 388.
Griscavage, J.M., et al. 1995. Biochem. Biophys. Res. Commun. 215, 721.
Squier, M.K., et al. 1994. J. Cell Physiol.159, 229.
Rami, J., and Kreiglstein, J. 1993. Brain Res.609, 67.
Sherwood, S.W., et al. 1993. Proc. Natl. Acad. Sci.USA90, 3353.
Vinitsky, A., et al. 1992. Biochemistry31, 9421.
Sasaki, T., et al. 1990. J. Enzyme Inhib. 3, 195.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Kai Wang et al.
Circulation, 145(9), 675-687 (2022-02-23)
High blood cholesterol accelerates the progression of atherosclerosis, which is an asymptomatic process lasting for decades. Rupture of atherosclerotic plaques induces thrombosis, which results in myocardial infarction or stroke. Lowering cholesterol levels is beneficial for preventing atherosclerotic cardiovascular disease. Low-density
J Michael Younger et al.
Methods in molecular biology (Clifton, N.J.), 301, 293-303 (2005-05-27)
Components of the ubiquitin-proteasome system function on the surface of the endoplasmic reticulum (ER) to select misfolded proteins for degradation. Herein we describe methods that allow for the study of the pathway for proteasomal degradation of the cystic fibrosis transmembrane
Abdallah Mound et al.
PloS one, 18(1), e0279028-e0279028 (2023-01-21)
Nod-Like Receptor Pyrin domain-containing protein 6 (NLRP6), a member of the Nucleotide-oligomerization domain-Like Receptor (NLR) family of proteins, assembles together with the ASC protein to form an inflammasome upon stimulation by bacterial lipoteichoic acid and double-stranded DNA. Besides its expression
Yoshinori Takeuchi et al.
iScience, 24(12), 103446-103446 (2022-01-07)
KLF15 is a transcription factor that plays an important role in the activation of gluconeogenesis from amino acids as well as the suppression of lipogenesis from glucose. Here we identified the transcription start site of liver-specific KLF15 transcript and showed
Yuhang Shi et al.
Viruses, 15(9) (2023-09-28)
SERINC5 is a restriction factor that becomes incorporated into nascent retroviral particles, impairing their ability to infect target cells. In turn, retroviruses have evolved countermeasures against SERINC5. For instance, the primate lentiviruses (HIV and SIV) use Nef, Moloney Murine Leukemia

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service