Skip to Content
Merck
All Photos(1)

Key Documents

20-148

Sigma-Aldrich

HAT Assay Buffer, 5X

For use in the HAT Assay.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (histone acetyltransferase (HAT))

shipped in

wet ice

Application

Research Category
All

Quality

Routinely evaluated in a HAT Assay

Physical form

250mM Tris-base, pH 8.0, 50% glycerol, 0.5mM EDTA and 5mM dithiothreitol

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of action of cyclosporin A on islet alpha- and beta-cells. Effects on cAMP- and calcium-dependent pathways.
F Martin,F J Bedoya
Life Sciences null
Preparation, characterization and 32P-postlabeling of butadiene monoepoxide N6-adenine adducts.
P Koivisto,R Kostiainen,I Kilpelainen,K Steinby,K Peltonen
Carcinogenesis null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service