Skip to Content
Merck
All Photos(1)

Key Documents

172426

Sigma-Aldrich

Glucoamylase, Rhizopus sp.

Glucoamylase is a multi-domain enzyme that belongs to the glycosyl hydrolase 15 family. Useful for the enzymatic determination of starch.

Synonym(s):

Glucoamylase, Rhizopus sp., Glucan 1,4-α-glucosidase, Glucoamylase, Amyloglucosidase, γ-Amylase, Lysosomal α-glucosidase, exo-4,7-α-glucosidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

fungus (Rhizopus spp.)

Quality Level

form

lyophilized

specific activity

≥10 unit/mg solid
≥20 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: 1 mg/mL
water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Native glucoamylase from Rhizopus species. Glucoamylase is a multi-domain enzyme that belongs to the glycosyl hydrolase 15 family. It catalyzes the hydrolysis of 1,4-linked α-D-glucose residues in glucosylpolysaccharides, liberating glucose units. Useful for the enzymatic determination of starch.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will liberate 1 µmol glucose from maltopentaose per min at 37°C, pH 6.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Hiromi, K., et al. 1973. Biochim. Biophys. Acta302, 362.
Kelly, J.J. and Alpers, D.H. 1973. Biochim. Biophys. Acta315, 113.
Jeffrey, P.L., et al. 1970. Biochemistry9, 1403.
Brown, B. and Brown, D.H. 1965. Biochim. Biophys. Acta110, 124.
Tsujisaka, Y., et al. 1958. Nature181, 770.
Miller, K.D. and Copeland, W.H. 1956. Biochim. Biophys. Acta22, 193.
French, D. and Knapp, D.W. 1950. J. Biol. Chem.187, 463.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service