Skip to Content
Merck
All Photos(1)

Key Documents

16-237

Sigma-Aldrich

Normal Rabbit IgG, Alexa Fluor 488 conjugate

Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunofluorescence.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

clone

polyclonal

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Fractionation and ion exchange chromatography

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P L Tazzari et al.
Leukemia, 21(3), 427-438 (2007-01-12)
A high incidence of relapses following induction chemotherapy is a major hindrance to patient survival in acute myelogenous leukemia (AML). There is strong evidence that activation of the phosphoinositide 3 kinase (PI3K)/Akt signaling network plays a significant role in rendering
C Welch et al.
Metabolism: clinical and experimental, 44(2), 234-238 (1995-02-01)
Recent studies have suggested that nitric oxide (NO) may function as both an intracellular and intercellular signal that affects neural and immunological activity, vascular tone, platelet adhesion, and production of some hormones. Arginine analogs such as NG-monomethyl-L-arginine (L-NMMA) and N
Hao Zhang et al.
Frontiers in plant science, 9, 1520-1520 (2018-11-22)
In plants, lateral roots play a crucial role in the uptake of water and nutrients. Several genes such as Zea mays Haem Oxygenase-1 (ZmHO-1) and Giberellic Acid-Stimulated Like-1 (ZmGSL-1) have been found to be involved in lateral root development. In
Shikha Nayar et al.
Nature, 593(7858), 275-281 (2021-04-01)
Crohn's disease is a chronic inflammatory intestinal disease that is frequently accompanied by aberrant healing and stricturing complications. Crosstalk between activated myeloid and stromal cells is critical in the pathogenicity of Crohn's disease1,2, and increases in intravasating monocytes are correlated
Pediatric early T-cell precursor leukemia with NF1 deletion and high-sensitivity in vitro to tipifarnib.
C Biagi et al.
Leukemia, 24(6), 1230-1233 (2010-04-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service