Skip to Content
Merck
All Photos(1)

Key Documents

16-189

Sigma-Aldrich

Anti-phospho-Histone H3 (Ser10) Antibody, biotin conjugate

Upstate®, from rabbit

Synonym(s):

H3 histone, family 3A, H3S10P, Histone H3 (phospho S10), H3 histone, family 3A, H3 histone, family 3B, H3 histone, family 3B (H3.3B)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... HIST1H3F(8968)

Immunogen

KLH-conjugated peptide (ARK[pS]TGGKAPRKQL- C) corresponding to amino acids 7-20 of human histone H3.

Application

Anti-phospho-Histone H3 (Ser10) Antibody, biotin conjugate is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10. This purified mAb, also known as H3S10p,is biotinylated, published in peer reviewed journals and has been validated in WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated in immunocytochemistry by immunostaining chromatin in HeLa cells undergoing mitosis

Physical form

PBS, pH 7.4, with 0.05% sodium azide.
Protein A purified

Storage and Stability

6 months at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An-Sofie Stevens et al.
Scientific reports, 7, 45616-45616 (2017-05-04)
One of the major challenges in the development of alternative carcinogenicity assays is the prediction of non-genotoxic carcinogens. The variety of non-genotoxic cancer pathways complicates the search for reliable parameters expressing their carcinogenicity. As non-genotoxic and genotoxic carcinogens have different
Jason D Gray et al.
Human molecular genetics, 22(21), 4267-4281 (2013-06-19)
Low-density lipoprotein receptor related protein 6 (Lrp6) mutational effects on neurulation were examined using gain (Crooked tail, Lrp6(Cd)) and loss (Lrp6(-)) of function mouse lines. Two features often associated with canonical Wnt signaling, dorsal-ventral patterning and proliferation, were no different
An-Sofie Stevens et al.
Journal of applied toxicology : JAT, 35(3), 319-326 (2014-06-27)
To investigate hydrophobic test compounds in toxicological studies, solvents like dimethylsulfoxide (DMSO) are inevitable. However, using these solvents, the interpretation of test compound-induced responses can be biased. DMSO concentration guidelines are available, but are mostly based on acute exposures involving
Jason D Gray et al.
Human molecular genetics, 19(23), 4560-4572 (2010-09-17)
Crooked tail (Cd) mice bear a gain-of-function mutation in Lrp6, a co-receptor for canonical WNT signaling, and are a model of neural tube defects (NTDs), preventable with dietary folic acid (FA) supplementation. Whether the FA response reflects a direct influence
An-Sofie Stevens et al.
Toxicological sciences : an official journal of the Society of Toxicology, 162(1), 251-263 (2017-11-18)
Aiming to in vivo characterize the responses of pluripotent stem cells and regenerative tissues to carcinogenic stress, we employed the highly regenerative organism Schmidtea mediterranea. Its broad regenerative capacities are attributable to a large pool of pluripotent stem cells, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service