Skip to Content
Merck
All Photos(1)

Key Documents

14-864

Sigma-Aldrich

PAK-1 PBD Protein (agarose free), 300 µg

For use in Affinity Binding Assays, ELISA & G-Protein Assays.

Synonym(s):

p21-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

mol wt

Mw 21 kDa

manufacturer/tradename

Upstate®

concentration

>50% (Coomassie blue staining, SDS-PAGE)
2 mg/mL

technique(s)

ELISA: suitable
affinity binding assay: suitable (G-protein)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

General description

Product Source: GST fusion-protein, corresponding to the p21-binding domain (PBD, residues 67-150) of human PAK-1, expressed in E. coli.

Quality

Routinely evaluated by G-protein activity assay

Physical form

300 µg of PAK-1 PBD, GST in 150 µL of PBS, pH 7.4, 0.5 mM PMSF, containing 20% glycerol.

Storage and Stability

Stable for 1 year at -20C from date of shipment. For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Specific Activity: Affinity Precipitation Assay: 10 µg of this lot when added with 10 µL glutathione agarose, precipitated Rac-GTP from 3T3 and A431 cell lysates that were loaded in vitro with GTPγS. GDP treated of 3T3 and A431 lysates served as negative controls. Precipitated Rac-GTP was detected by immunoblot analysis using 1 &micor;g/mL anti-Rac (Catalog # 05-389) and secondary goat anti-mouse HRP antibody (Catalog # 12-349) on a Western Blot.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A X Bigard et al.
European journal of applied physiology and occupational physiology, 76(5), 421-427 (1997-01-01)
We examined the extent of morphological alterations and the myosin heavy chain (MHC) distribution in the rat soleus muscle after a 4-week period of spontaneous recovery or retraining after hindlimb suspension (HS). Moreover, we tested the hypothesis that dantrolene sodium
A Ros-Baro et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(21), 12050-12055 (2001-10-11)
It has been recently reported that insulin recruits a novel signaling machinery to lipid rafts required for insulin-stimulated GLUT4 translocation [Baumann, A., Ribon, V., Kanzaki, M., Thurmond, D. C., Mora, S., Shigematsu, S., Bickel, P. E., Pessin, J. E. &
Zhenyuan Xu et al.
International journal of molecular sciences, 22(9) (2021-06-03)
Schwann cells (SCs) are a highly plastic cell type capable of undergoing phenotypic changes following injury or disease. SCs are able to upregulate genes associated with nerve regeneration and ultimately achieve functional recovery. During the regeneration process, the extracellular matrix
Shamsideen A Ojelade et al.
PloS one, 10(9), e0137465-e0137465 (2015-09-15)
Responses to the effects of ethanol are highly conserved across organisms, with reduced responses to the sedating effects of ethanol being predictive of increased risk for human alcohol dependence. Previously, we described that regulators of actin dynamics, such as the
Tudor I Oprea et al.
PloS one, 10(11), e0142182-e0142182 (2015-11-13)
Rho family GTPases (including Rac, Rho and Cdc42) collectively control cell proliferation, adhesion and migration and are of interest as functional therapeutic targets in numerous epithelial cancers. Based on high throughput screening of the Prestwick Chemical Library® and cheminformatics we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service