Skip to Content
Merck
All Photos(1)

Key Documents

1.46151

Millipore

Malt Extract Agar

Ready-to-use 90mm plate for detection of yeasts and moulds by Membrane filtration method.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

Quality Level

form

solid

feature

ready-to-use

packaging

pkg of (90 mm plate (20 plates per box))

pH

5.6 ( in H2O)

application(s)

air monitoring
food and beverages
industrial qc
microbiology

suitability

molds
yeasts

General description

Malt Extract Agar plates are stackable specially designed plates that helps increase the safety during handling, transport and incubation and are of standard 90mm diameter. Malt Extract Agar is used for the detection of yeasts and moulds by Membrane filtration method. The pH of 5.4 - 5.8 of the medium promotes the growth of most yeasts and fungi, while bacterial growth is strongly inhibited. Malt extract has a high content of carbohydrates (90-92 %), nitrogen compounds and vitamins, e.g. riboflavin, pyridoxin, and thiamine. The soymeal peptone is also rich in carbohydrates, and additionally supplies the necessary protein compounds such as peptides and amino acids.

Application

Malt Extract Agar is used for the identification of yeasts and molds in food, including milk and dairy products, and also in environmental samples. It is particularly recommended for the analysis of beverages in which yeasts are a predominant ingredient.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service