Skip to Content
Merck
All Photos(2)

Key Documents

14-404-M

Sigma-Aldrich

ZAP-70 Protein, active, 20 µg

Active, recombinant, human, full length ZAP-70 containing a C-terminal His6 tag, for use in Kinase Assays.

Synonym(s):

70 kDa zeta-chain associated protein, SRK, Syk-related tyrosine kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

form

liquid

shelf life

6 mo.

mol wt

Mw 70 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ZAP70(7535)

General description

Research area: CELL SIGNALING

Product Source: expressed in Sf21 Recombinant, human, full length ZAP-70 containing a C-terminal His6-tag

Zeta-chain-associated protein kinase 70 (ZAP-70) is a cytoplasmic tyrosine kinase that consists of tandem SH2 (tSH2) domains at the beginning, linked with a long linker region, and a tyrosine kinase domain at the C-terminal end.

Biochem/physiol Actions

Protein Target: ZAP-70

Target Sub-Family: TK

Zeta-chain-associated protein kinase 70 (ZAP-70) plays a vital role in the development and function of T cells and natural killer cells. It interacts directly with the activated T-cell receptor and regulates downstream signals. In addition, ZAP-70 also mediates B-cell receptor (BCR) signaling. Mutations in ZAP-70 have been associated with severe combined immunodeficiency (SCID), chronic lymphocytic leukemia (CLL) , and the development of autoreactive T cells that attack the joints.

Quality

routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1) substrate peptide

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -20C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Structural Basis for Activation and Inhibition of ZAP-70 Kinase Domain
Huber RG, et al.
PLoS Computational Biology, 11(10), e1004560-e1004560 (2015)
ZAP-70: an essential kinase in T-cell signaling
Wang H, et al.
Cold Spring Harbor Perspectives in Biology, 2(5), a002279-a002279 (2010)
Experimental Models for Rheumatoid Arthritis
Rikard Holmdahl
Journal of Separation Science, 400-412 (2013)
ZAP-70 in Signaling, Biology, and Disease
Au-Yeung BB, et al.
Annual Review of Immunology, 36, 127-156 (2018)
D H Chu et al.
Immunological reviews, 165, 167-180 (1998-12-16)
The processes of T-cell development and activation employ similar immature and mature receptors as well as similar signal transduction pathways to achieve different outcomes. Many signaling molecules are shared between the receptor signaling pathways, including two families of cytoplasmic protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service