Skip to Content
Merck
All Photos(1)

Key Documents

126609

Sigma-Aldrich

Albumin, Bovine Serum, Fraction V, Fatty Acid-Free, Nuclease- and Protease-Free

BSA Fatty Acid-free is designed for use in serological testing, RIA, and hormone response studies. Suitable for use in Molecular Biology applications, such as Northern and Southern blots.

Synonym(s):

Albumin, Bovine Serum, Fraction V, Fatty Acid-Free, Nuclease- and Protease-Free

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

description

Merck USA index - 14, 8468

Assay

≥98% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.02% fatty acids
≤3.0% Ash
≤5% moisture

color

off-white to beige

solubility

0.15 M sodium chloride: 10 mg/mL
water: 10 mg/mL

cation traces

heavy metals: ≤10 ppm

foreign activity

endonuclease, none detected
protease ≤0.1 U/mg

shipped in

ambient

storage temp.

2-8°C

General description

Designed for use in serological testing, RIA, and hormone response studies. Suitable for use in Molecular Biology applications, such as Northern and Southern blots. Also useful as a reagent protein standard and as a nutrient in tissue culture where a higher purity BSA is needed.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). 0.1% NaN₃ may be added as a preservative. Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Thomassen, D.G. 1989. In Vitro Cell. Dev. Biol.25, 1046.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daan P Hurkmans et al.
Journal for immunotherapy of cancer, 8(2) (2021-01-12)
Many cancer patients do not obtain clinical benefit from immune checkpoint inhibition. Checkpoint blockade targets T cells, suggesting that tyrosine kinase activity profiling of baseline peripheral blood mononuclear cells may predict clinical outcome. Here a total of 160 patients with
Roozbeh Khosravi et al.
Bone, 58, 33-41 (2013-10-15)
Diabetes increases the risk of bone fracture. Organic and inorganic bone extracellular matrix components determine bone strength. Previous studies indicate that in diabetes, glycation of collagen causes abnormal arrangements of collagen molecules and fragile bones. Diabetic bone fragility is additionally
Eric E Niederkofler et al.
PloS one, 8(11), e81125-e81125 (2013-11-28)
Insulin-like growth factor 1 (IGF1) is an important biomarker of human growth disorders that is routinely analyzed in clinical laboratories. Mass spectrometry-based workflows offer a viable alternative to standard IGF1 immunoassays, which utilize various pre-analytical preparation strategies. In this work
Atsuko Uchida et al.
Methods in cell biology, 131, 21-90 (2016-01-23)
Neurofilaments, which are the intermediate filaments of nerve cells, are space-filling cytoskeletal polymers that contribute to the growth of axonal caliber. In addition to their structural role, neurofilaments are cargos of axonal transport that move along microtubule tracks in a
Cindy Hodakoski et al.
Cancers, 11(1) (2019-01-06)
Cancer cells can adapt to nutrient poor conditions by rewiring their metabolism and using alternate fuel sources. Identifying these adaptive metabolic pathways may provide novel targets for cancer therapy. Here, we identify a subset of non-small cell lung cancer (NSCLC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service