Skip to Content
Merck
All Photos(1)

Key Documents

124029

Sigma-Aldrich

Akt Inhibitor XII, Isozyme-Selective, Akti-2

The Akt Inhibitor XII, Isozyme-Selective, Akti-2 controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor XII, Isozyme-Selective, Akti-2, 1-((2-(4-((4-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl)methyl)phenyl)-3-phenyl-6-quinoxalinyl)carbonyl)-4-methyl-piperazine, 3HCl and, 1-((3-(4-((4-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl)methyl)phenyl)-2-phenyl-6-quinoxalinyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H39N7O2 · 3HCl
Molecular Weight:
747.16
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (mixture of regioisomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 25 mg/mL
water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable Akti-1/2- (Cat. No. 124018) derived allosteric inhibitor pair (1:1 regioisomeric mixture) with much improved aqueous solubility (10 mg/ml in a pH 7.4 saline) and Akt2 selectivity (IC50 = 805 nM for Akt2, >10 µM for Akt1/3, and >50 µM for PKA/PKC/SGK). The inhibition is PH domain-dependent and non-competitive with ATP.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth K Y Ting et al.
ImmunoHorizons, 8(1), 57-73 (2024-01-09)
The accumulation of lipid and the formation of macrophage foam cells is a hallmark of atherosclerosis, a chronic inflammatory disease. To better understand the role of macrophage lipid accumulation in inflammation during atherogenesis, we studied early molecular events that follow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service