Skip to Content
Merck
All Photos(1)

Key Documents

1.19806

Supelco

Zinc standard solution

traceable to SRM from NIST Zn(NO₃)₂ in HNO₃ 0.5 mol/l 1000 mg/l Zn Certipur®

Synonym(s):

Zinc Reference Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

product line

Certipur®

concentration

1000 mg/L Zn in 0.5 M HNO3

technique(s)

AAS: suitable

pH

0.48 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Zinc standard solution may be used as a calibration standard in the determination of zinc in pure copper and nickel-based alloy by electrothermal atomic absorption spectrometry.[1]

Analysis Note

1000 mg/L Zn in 0.5 mol/L HNO3, prepared with high-purityZn(NO3)2, HNO3, and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binary modifiers for the determination of zinc in pure copper and nickel-based alloy by longitudinal Zeeman electrothermal atomic absorption spectrometry
Chen S, et al.
Analytica Chimica Acta, 550, 156-163 (2005)
Mojtaba Abdollahi et al.
Archives of animal nutrition, 74(3), 189-205 (2019-12-19)
This study was conducted to assess the effect of feeding high-surface ZnO instead of common ZnO on the performance, rumen fermentation, blood minerals, leukocytes and antioxidant capacity of pre- and post-weaning calves. Thirty male suckling Holstein calves were allotted to

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service