1.15165
Amberlite™ MB-6113 Ion Exchange Resin
matrix mixed-bed, 15-50 mesh
Synonym(s):
polymeric adsorbent
About This Item
Recommended Products
Product Name
Ion exchanger Amberlite™ MB-6113, suitable for ion chromatography, matrix (strongly acidic H+form and strongly basic OH- form)
form
resin
Quality Level
technique(s)
HPLC: suitable
LPLC: suitable
ion chromatography: suitable
matrix
(strongly acidic H+form and strongly basic OH- form)
PS-DVB
particle size
355-1180 μm
separation technique
mixed mode
storage temp.
15-25°C
Related Categories
General description
Analysis Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service