Skip to Content
Merck
All Photos(1)

Key Documents

1.13741

Sigma-Aldrich

Lactophenol blue solution

for staining fungi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.74

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

2.3 (20 °C in H2O)

density

1.16 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Lactophenol blue solution - for staining fungi is a ready-to-use staining solution for fungi staining in bacteriology with samples of human origin. The material is stained in one step with Lactophenol blue solution. Fungi appear blue. This ready-to-use solution is especially for air-dried, heat-fixed smears of bacteriological material, e.g. sputum, FNAB, lavages, imprints, body fluids, puss, and exudates, liquid and solid cultures.The staining solution is sufficient for up to 100 preparations per bottle. It is developed and registered as CE / IVD product for applications in diagnostics. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
Yeasts / Moulds: blue

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service