Skip to Content
Merck
All Photos(3)

Key Documents

1.12636

Supelco

Acetonitrile

for DNA synthesis (max. 10 ppm H2O)

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-835-2
NACRES:
NA.22

grade

anhydrous

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

524 °C
973 °F

expl. lim.

16 %

technique(s)

DNA amplification: suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Additional to our high quality acetonitrile dried with low water content up to 10 ppm our DNA-/RNA synthesis reagents with its high quality, lowest water and acid content are best requirements for a reliable and reproducible DNA-/RNA synthesis with high yields. A wide variety of packaging types supporting specific synthesis instrumentations, for quantities from bottle to bulk, and the will to customized solutions completes our innovative offering.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service