1.11723
Casein Digest Soy Lecithin Polysorbate 20 (FCDLP 20) Broth (Base)
NutriSelect® Prime, according to ISO 16212, ISO 18415, ISO 21149 (TAT Broth Base), powder, pack of 500 g
Synonym(s):
TAT broth
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
Recommended Products
Agency
ISO 18415
ISO 21149 (TAT Broth Base)
according to ISO 16212
Quality Level
description
For sterility tests of viscous or gelatinous materials
sterility
non-sterile
form
powder
packaging
pack of 500 g
manufacturer/tradename
NutriSelect® Prime
technique(s)
microbiological culture: suitable
pH
7.3±± 0.2
application(s)
cosmetics
microbiology
pharmaceutical
storage temp.
15-25°C
Related Categories
General description
Casein-peptone lecithin polysorbate broth (base) is a powdered medium recommended for general microbiological analysis. Casein enzymic hydrolysate provides the nitrogen, vitamins, amino acids, and carbon in the broth base. Casein peptone in the medium provides optimal conditions for the spore germination and regeneration of the damaged microorganisms. Lecithin and polysorbate 20 inactivate many antimicrobial compounds.
Application
Casein-peptone lecithin polysorbate broth (base) is recommended for diluting samples of pharmaceutical, cosmetic, and other raw materials or final products for determining the total microbial counts.
Analysis Note
- Appearance (clearness): clear to almost clear
- Appearance (colour): yellowish
- pH-value (25 °C): 7.1 - 7.5
- Growth (Staphylococcus aureus ATCC 6538 (WDCM 00032)): 91%
- Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): 77%
- Growth (Escherichia coli ATCC 8739 (WDCM 00012)): 101%
- Growth (Candida albicans ATCC 10231 (WDCM 00054)): 72%
- Incubation: 45 minutes up to 1 hour at 18 – 27°C (laboratory ambient temperature), aerobic
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service