1.10989
Pseudomonas agar F (base)
for microbiology
Synonym(s):
Pseudomonas agar F
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
Recommended Products
Quality Level
form
medium granules (dehydrated (DCM))
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
pH
7.1-7.3 (25 °C, 35 g/L in H2O, after autoclaving)
solubility
35 g/L
bulk density
610 kg/m3
application(s)
food and beverages
microbiology
storage temp.
15-25°C
suitability
Pseudomonas spp.
Related Categories
Analysis Note
Appearance (clearness): claer to turbid
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.0 - 7.4
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): good to very good
Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): good to very good
Growth (Pseudomonas fluorescens ATCC 17397): good to very good
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)): good to very good
Yellow-green pigment (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): +
Yellow-green pigment (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Yellow-green pigment (Pseudomonas fluorescens ATCC 17397): +/-
Yellow-green pigment (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Yellow-green pigment (Escherichia coli ATCC 25922 (WDCM 00013)): -
Yellow-green pigment (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Fluorescence (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): +
Fluorescence (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Fluorescence (Pseudomonas fluorescens ATCC 17397): +/-
Fluorescence (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Fluorescence (Escherichia coli ATCC 25922 (WDCM 00013)): -
Fluorescence (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Incubation: 24 hrs. (Pseudomonas fluorescens ATCC 17397 48 hrs.); 35 °C; aerobic.
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.0 - 7.4
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): good to very good
Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): good to very good
Growth (Pseudomonas fluorescens ATCC 17397): good to very good
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Enterobacter cloacae ATCC 13047 (WDCM 00083)): good to very good
Yellow-green pigment (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): +
Yellow-green pigment (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Yellow-green pigment (Pseudomonas fluorescens ATCC 17397): +/-
Yellow-green pigment (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Yellow-green pigment (Escherichia coli ATCC 25922 (WDCM 00013)): -
Yellow-green pigment (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Fluorescence (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): +
Fluorescence (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Fluorescence (Pseudomonas fluorescens ATCC 17397): +/-
Fluorescence (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): -
Fluorescence (Escherichia coli ATCC 25922 (WDCM 00013)): -
Fluorescence (Enterobacter cloacae ATCC 13047 (WDCM 00083)): -
Incubation: 24 hrs. (Pseudomonas fluorescens ATCC 17397 48 hrs.); 35 °C; aerobic.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service