1.10864
GranuCult® prime Iron sulfite Agar (base)
according to ISO ISO 15213-1
Synonym(s):
ISA, Sulfite iron agar (SIA), Tryptose sulfite cycloserine agar
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
Recommended Products
Agency
according to ISO
Quality Level
description
for microbiology
pH
6.7-7.1 ( in H2O)
storage temp.
15-25°C
Analysis Note
Appearance (clearness): clear
Appearance (colour): yellowis to yellow-green
pH-value (25 °C): 6.7 - 7.1
Growth (Clostridium perfringens ATCC 10543): good to very good
Growth (Clostridium perfringens ATCC 13124 (WDCM 00007)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): fair to very good
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): poor to good
Growth (Bacillus cereus ATCC 11778 (WDCM 00001)): poor to good
Black colonies (Clostridium perfringens ATCC 10543): +
Black colonies (Clostridium perfringens ATCC 13124 (WDCM 00007)): +
Black colonies (Escherichia coli ATCC 25922 (WDCM 00013)): -
Black colonies (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Black colonies (Bacillus cereus ATCC 11778 (WDCM 00001)): -
Incubation: 24 hrs.; 35 °C; aerobic.
Appearance (colour): yellowis to yellow-green
pH-value (25 °C): 6.7 - 7.1
Growth (Clostridium perfringens ATCC 10543): good to very good
Growth (Clostridium perfringens ATCC 13124 (WDCM 00007)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): fair to very good
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): poor to good
Growth (Bacillus cereus ATCC 11778 (WDCM 00001)): poor to good
Black colonies (Clostridium perfringens ATCC 10543): +
Black colonies (Clostridium perfringens ATCC 13124 (WDCM 00007)): +
Black colonies (Escherichia coli ATCC 25922 (WDCM 00013)): -
Black colonies (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Black colonies (Bacillus cereus ATCC 11778 (WDCM 00001)): -
Incubation: 24 hrs.; 35 °C; aerobic.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service