Skip to Content
Merck
All Photos(2)

Key Documents

1.09940

Supelco

Sodium carbonate solution

c(Na₂CO₃) = 0.05 mol/l (0.1 N), concentrate for preparing 1000 ml volumetric solution for titration, Titrisol®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

product name

Sodium carbonate solution, for 1000 ml, c(Na₂CO₃) = 0.05 mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

concentration

0.05 M

technique(s)

titration: suitable

pH

11.6 (20 °C in H2O)

density

1.1 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service