1.09885
Buffer concentrate
(citrate/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 5.00 ± 0.02 (20°C) Titrisol®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161704
NACRES:
NA.25
Recommended Products
grade
analytical standard
Quality Level
Agency
according to DIN 19266
product line
Titrisol®
pH
5.00 (20 °C in H2O)
density
1.14 g/cm3 at 20 °C
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
Related Categories
General description
The Titrisol® reference buffer concentrate [citrate/sodium hydroxide; pH=5.00±0.02 (20 °C)] is analyzed in the ISO/IEC 17025 accredited calibration laboratory with a combined glass electrode and 5-point calibration according to DIN 19268 against a certified primary standard reference material following DIN 19266.
The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.
The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.
Features and Benefits
- Traceable to primary standard reference materials from NIST and PTB
- Analytical standard Grade
- Uncertainty data and traceability are documented in a comprehensive and batch-specific Certificate of Analysis.
Analysis Note
pH-value (20 °C): 5.00 +/- 0.02
Measured after dilution to 500 ml with a combination glass electrode after multiple point calibration (according to DIN 19268) with reference materials for the preparation of standard buffer solutions according to DIN 19266.
Measured after dilution to 500 ml with a combination glass electrode after multiple point calibration (according to DIN 19268) with reference materials for the preparation of standard buffer solutions according to DIN 19266.
Legal Information
Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany
standard
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service