1.09492
ICP multi-element standard solution VIII
(24 elements in dilute nitric acid) 100 mg/l: Al, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Se, Sr, Te, Tl, Zn Certipur®
Synonym(s):
Nota 1
About This Item
Recommended Products
grade
reference material
Quality Level
product line
Certipur®
technique(s)
ICP: suitable
pH
1 (20 °C in H2O)
bp
100 °C/1013 hPa
density
1.039 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
multi-component solution
storage temp.
15-25°C
Analysis Note
B (boron): 100 mg/l
Ba (barium): 100 mg/l
Be (beryllium): 100 mg/l
Bi (bismuth): 100 mg/l
Ca (calcium): 100 mg/l
Cd (cadmium): 100 mg/l
Co (cobalt): 100 mg/l
Cr (chromium): 100 mg/l
Cu (Copper): 100 mg/l
Fe (iron): 100 mg/l
Ga (Gallium): 100 mg/l
K (potassium): 100 mg/l
Li (Lithium): 100 mg/l
Mg (Magnesium): 100 mg/l
Mn (Manganese): 100 mg/l
Na (sodium): 100 mg/l
Ni (Nickel): 100 mg/l
Pb (lead): 100 mg/l
Se (Selenium): 100 mg/l
Sr (Strontium): 100 mg/l
Te (Tellurium): 100 mg/l
Tl (Thallium): 100 mg/l
Zn (Zinc): 100 mg/l
Other Notes
Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1
Supplementary Hazards
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service