Skip to Content
Merck
All Photos(1)

Key Documents

1.08446

Supelco

Titriplex® III solution

c(Na₂-EDTA 2 H₂O) = 0.01 mol/l, concentrate for preparing 1000 ml volumetric solution for titration, Titrisol®

Synonym(s):

EDTA solution ; Ethylenediaminetetraacetic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.21

product name

Titriplex® III solution, for 1000 ml, c(Na₂-EDTA 2 H₂O) = 0.01 mol/l Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Complexometric reactions
reaction type: Redox Reactions

concentration

0.01 M

technique(s)

titration: suitable

pH

7.8 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

storage temp.

15-25°C

General description

for 1000 ml, c(Na₂-EDTA 2 H₂O) = 0.01 mol/l Titrisol®

Analysis Note

Concentration after dilution to 1 liter: c(C₁₀H₁₄N₂Na₂O₈ ∙ 2 H₂O) = 0.01 mol/lAmount-of-substance concentration0.01000 mol/lThe concentration of this solution was determined with volumetric standard zinc (article number 1.02409). The determined titer at 20°C was 1.000 with an expanded measurement uncertainty of ± 0.004 (k=2 coverage factor for 95% coverage probability). The certified value is traceable to primary standard NIST SRM 682 (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard zinc, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany
Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service