Skip to Content
Merck
All Photos(1)

Key Documents

1.07299

Sigma-Aldrich

4-(Methylamino)phenol sulfate

Technipur®

Synonym(s):

4-(Methylamino)phenol hemisulfate salt, N-Methyl-p-aminophenol sulphate, p-Methyl aminophenol sulfate, Metol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H9NO · 1/2H2SO4
CAS Number:
Molecular Weight:
172.19
Beilstein:
3919382
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.05

Quality Level

autoignition temp.

989 °F

pH

3.5-4.5 (20 °C, 50 g/L in H2O)

mp

260 °C (dec.) (lit.)

SMILES string

OS(O)(=O)=O.CNc1ccc(O)cc1.CNc2ccc(O)cc2

InChI

1S/2C7H9NO.H2O4S/c2*1-8-6-2-4-7(9)5-3-6;1-5(2,3)4/h2*2-5,8-9H,1H3;(H2,1,2,3,4)

InChI key

ZVNPWFOVUDMGRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Methylamino)Phenol Sulfate, a substituted phenol, is an aromatic reducing agent, majorly used as a hair dye and a monochrome photographic developer chemical.

Application

4-(Methylamino)Phenol Sulfate can be used as a corrosion inhibitor. It is used as a reagent in several reactions employed to determine the levels of contaminants and drugs by various means, particularly by electrochemical or spectrophotometric methods. 4-(Methylamino)Phenol Sulfate is also used as an organic reducing agent to prepare silver nanomaterials.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

494.6 °F - closed cup

Flash Point(C)

257 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Innovation of Novel Stone-Like Perovskite Structured Calcium
Stannate (CaSnO3): Synthesis, Characterization, and Application
Headed for Sensing Photographic Developing Agent Metol
Muthukutty B, et al.
ACS sustainable chemistry & engineering, 8(11), 4419-4430 (2020)
Metol as Corrosion Inhibitor for Steel
Praveen BM and Venkatesha TV
International Journal of Electrochemical Science, 4 , 267-275 (2009)
ZrO2 supported Nano-ZSM-5 nanocomposite material for the nanomolar electrochemical detection of metol and bisphenol A
Kaur B, et al.
Royal Society of Chemistry Advances, 6, 65736-65746 (2016)
Correction to Active-Site-Rich 1T-Phase CoMoSe2 Integrated Graphene Oxide Nanocomposite as an Efficient Electrocatalyst for Electrochemical Sensor and Energy Storage Applications
Ramaraj S, et al.
Analytical Chemistry, 92(2), 2347-2348 (2020)
Catalytic Determination of Vanadium Based on the
Bromate Oxidative-Coupling Reaction
of Metol with Phloroglucinol
Mohamed AA and Fawy KF
Mikrochimica Acta, 134, 229-235 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service