Skip to Content
Merck

1.06839

Supelco

Sodalime pellets with indicator

for analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352319
NACRES:
NB.21

Pricing and availability is not currently available.

Quality Level

loss

13.0 - 18.0% loss on drying, (105°C)

color

, Passes test

particle size

2-5 mm, coeff var ≥95%

capacity

≥25 % absorption capacity (CO2)((method NF XVII))

bulk density

75 g/100 mL

suitability

passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service