Skip to Content
Merck

106333

SAFC

CellPrime® r LR3 IGF powder

Pharma Manufacturing

Synonym(s):

CellPrime® R3 IGF-I human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C400H619N111O115S9
CAS Number:
Molecular Weight:
9111.45
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

application(s)

pharma/biopharma processes
pharmaceutical

Quality Level

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

CellPrime® rLR3 IGF is a is a non-animal origin (NAO) recombinant equivalent Insulin-like growth factor expressed in E.coli. CellPrime® rLR3 IGF has a 13 amino acid N-terminal extension and position 3 substitution of Glutamic acid to Arginine contributes to increased stability and robustness in media formulations.

Application

CellPrime® rLR3 IGF is used to optimize cell culture formulations to increase cell proliferation, productivity, and viability. With an exact match for the Type I IGF receptor and a lower affinity for IGF binding proteins, CellPrime® rLR3 IGF has increased effectiveness in serum-free cell culture media. Concentrations required to enhance performance is unique to each cell line and may be optimized by using in combination with CellPrime® rInsulin.

Packaging

106333.0005 / 5 MG /
106333.0050/ 50 MG /
106333.0250/ 250 MG /
106333.0500 / 500 MG /

Legal Information

CellPrime is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service