Skip to Content
Merck
All Photos(2)

Key Documents

1.05986

Supelco

Marble granular

for producing CO₂

Synonym(s):

Calcium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
11111605
EC Index Number:
207-439-9
NACRES:
NB.21
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

form

solid

potency

6450 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

9.5-10.5 (20 °C, 100 g/L in H2O, slurry)

mp

825 °C (decomposition)

density

2.93 g/mL at 25 °C (lit.)

bulk density

300‑1400 kg/m3

storage temp.

no temp limit

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Identity: passes test
Particle size (0.5-2 mm): ≥ 95 %

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service