Skip to Content
Merck
All Photos(2)

Key Documents

1.05589

Supelco

Sodium hydroxide solution

30%, Suprapur®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Sodium hydroxide solution 30%, Suprapur®

grade

for inorganic trace analysis

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320
suitable for SM 5210

vapor pressure

3 mmHg ( 37 °C)

form

liquid

concentration

30%

pH

14 (20 °C in H2O)

density

1.33 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 30.0 %
Chloride (Cl): ≤ 1000 ppb
Phosphate (PO₄): ≤ 50 ppb
Silicate (as SiO₂): ≤ 2000 ppb
Sulfate (SO₄): ≤ 2000 ppb
Al (Aluminium): ≤ 20 ppb
As (Arsenic): ≤ 50 ppb
Ba (Barium): ≤ 200 ppb
Ca (Calcium): ≤ 100 ppb
Cd (Cadmium): ≤ 2 ppb
Ce (Cerium): ≤ 5 ppb
Co (Cobalt): ≤ 2 ppb
Cr (Chromium): ≤ 10 ppb
Cu (Copper): ≤ 5 ppb
Eu (Europium): ≤ 5 ppb
Fe (Iron): ≤ 10 ppb
Hg (Mercury): ≤ 10 ppb
K (Potassium): ≤ 2000 ppb
La (Lanthanum): ≤ 5 ppb
Li (Lithium): ≤ 200 ppb
Mg (Magnesium): ≤ 50 ppb
Mn (Manganese): ≤ 2 ppb
Ni (Nickel): ≤ 5 ppb
Pb (Lead): ≤ 10 ppb
Sc (Scandium): ≤ 5 ppb
Sm (Samarium): ≤ 5 ppb
Sr (Strontium): ≤ 100 ppb
Tl (Thallium): ≤ 5 ppb
Y (Yttrium): ≤ 5 ppb
Yb (Ytterbium): ≤ 5 ppb
Zn (Zinc): ≤ 10 ppb

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service