1.05448
Wort agar
GranuCult® plus, suitable for microbiology, for yeasts, for molds
Synonym(s):
Selective media for yeasts and molds, Yeasts and molds medium
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
granular
packaging
pkg of 5 kg
pkg of 500 g
manufacturer/tradename
GranuCult® plus
storage condition
dry media (Store tightly closed)
technique(s)
microbiological culture: suitable
color
beige
pH
5.0 (30 °C, 55 g/L in H2O, after autoclaving)
solubility
55 g/L
bulk density
520 kg/m3
application(s)
food and beverages
microbiology
storage temp.
15-25°C
suitability
molds
yeasts
Related Categories
General description
Application
Features and Benefits
- GranuCult® offers superior granulated culture media
- Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
- Excellent wettability, solubility, and free-flowing properties
- Convenient, with minimal component separation and clumping, even under warm or humid conditions
- High batch-to-batch reproducibility
- Prolonged shelf life of up to five years•High number of test strains exceeding all regulatory demands
- Granulation technology allows many supplements to be included, with no need to add these separately
Analysis Note
Appearance (colour): brownish
Solidification behaviour (2 hrs., 45 °C): liquid
pH-value (25 °C): 4.8 - 5.2
Recovery on test medium (Candida albicans ATCC 10231 (WDCM 00054)): ≥ 70 %
Recovery on test medium (Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)): ≥ 70 %
Recovery on test medium (Saccharomyces cerevisiae ATCC 9080): ≥ 70 %
Recovery on test medium (Rhodotorula mucilaginosa DSM 70403): ≥ 70 %
Growth (Geotrichum candidum DSM 1240): good to very good
Growth (Penicillium commune ATCC 10428): fair to very good
Growth (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): good to very good
Growth (Trichophyton ajelloi ATCC 28454): fair to good
Incubation: 7 days; 28 °C; aerobic (yeasts 3 days).
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service