Skip to Content
Merck
All Photos(1)

Key Documents

1.05267

Millipore

GranuCult® Mannitol egg yolk polymyxin agar (base)

acc. ISO 7932, ISO 21871, FDA-BAM, and APHA, suitable for microbiology

Synonym(s):

MYP agar, Bacillus Cereus Selective Agar, Cereus Selective Agar Base, MYEP agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA
ISO 21871
ISO 7932

Quality Level

reg. compliance

FDA (BAM)

sterility

non-sterile

form

granular

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (25 °C, 43 g/L in H2O, after autoclaving)

solubility

43 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Bacillus cereus

General description

GranuCult® Mannitol egg yolk polymyxin agar (base) is used for the enumeration of Bacillus cereus. This medium contains two indicator systems, mannitol, and phenol red and egg yolk. Enzymatic digest of casein and meat extract provide carbon and nitrogen sources. The selective inhibitory component polymyxin suppresses the growth of many unwanted microorganisms. Bacteria that ferment mannitol to produce acids produce yellow staining of the medium with phenol red as a pH indicator. Bacillus cereus does not ferment mannitol so the medium around the colonies remains unchanged or is discolored due to light alkalization by forming pink colonies. The lecithin present in egg yolk is cleaved by the Bacillus cereus-lecithinase, which leads to the formation of an opalescent white precipitation zone surrounding the pink colonies. Bacillus anthracis, Bacillus mycoides, Bacillus pseudodomycoides, Bacillus thuringiensis, and Bacillus weihenstephanensis may not be distinguishable from Bacillus cereus on this medium.

Application

GranuCult® Mannitol egg yolk polymyxin agar (base) is used for the enumeration of presumptive Bacillus cereus from food and animal feed as well as from environmental samples and other materials.

Analysis Note

Appearance (clearness): clear to slightly opalescent
Appearance (color): red
pH-value (25 °C): 7.0 - 7.4
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Bacillus cereus ATCC 11778 (WDCM 00001)):
Colony count (Bacillus cereus ATCC 11778 (WDCM 00001)):
Recovery on test medium (Bacillus cereus ATCC 11778 (WDCM 00001)): ≥ 50 %
Growth (Bacillus subtilis ATCC 6633 (WDCM 00003)): no limit
Growth (Escherichia coli ATCC 8739 (WDCM 00012)): total inhibition
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): total inhibition
Colonies (Bacillus cereus ATCC 11778 (WDCM 00001)): pink colonies with precipitation halo
Colonies (Bacillus subtilis ATCC 6633 (WDCM 00003)): yellow colonies without precipitation halo
Incubation:
B.cereus 24 ± 3 to 44 ± 4 hours at 30 ± 1 °C
other teststrains 44 ± 4 hours at 30 ± 1 °C
A recovery rate of 50 % is equivalent to a productivity value of 0.5.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service