1.05221
Dowex 50W-X8 Ion Exchange Resin
hydrogen form, 60-170 mesh, strongly acidic, cationic
About This Item
Recommended Products
Product Name
Ion exchanger Dowex 50W-X8, (strongly acidic cation exchanger, H⁺ form)
Quality Level
form
resin
autoignition temp.
>500 °C
technique(s)
HPLC: suitable
LPLC: suitable
matrix
PS-DVB
matrix active group
SCX
particle size
90-250 μm
capacity
640 - 720 g/kg water absorbing capacity
density
0.800 g/cm3 at 20 °C
separation technique
cation exchange
storage temp.
15-25°C
SMILES string
C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C
InChI
1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2
InChI key
NWUYHJFMYQTDRP-UHFFFAOYSA-N
Related Categories
Analysis Note
Particle size (0,09-0,25 mm): ≥ 70.0 %
Total capacity: ≥ 1.7 mol/l
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service