Skip to Content
Merck
All Photos(1)

Key Documents

1.05076

Supelco

Potassium perchlorate

for analysis EMSURE® ACS

Synonym(s):

Potassium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClKO4
CAS Number:
Molecular Weight:
138.55
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-912-9
NACRES:
NA.21
Assay:
99.5-100.5% (alkalimetric)
grade:
ACS reagent
form:
solid
solubility:
17 g/L

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.5-100.5% (alkalimetric)

form

solid

impurities

≤0.005% Insoluble matter

pH

5.0 (25 °C, 10 g/L in H2O)

mp

610 °C (decomposition)

solubility

17 g/L

bulk density

1150 kg/m3

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.001%

cation traces

Ca: ≤0.005%
Fe: ≤0.0005%
N: ≤0.0005%
Na: ≤0.005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[Cl-](=O)(=O)(=O)=O

InChI

1S/ClHO4.K/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

YLMGFJXSLBMXHK-UHFFFAOYSA-M

Related Categories

Application


  • Temperature-Induced Change of Water Structure in Aqueous Solutions of Some Kosmotropic and Chaotropic Salts.: Investigates how temperature variations alter the water structure in solutions containing kosmotropic and chaotropic salts, including potassium perchlorate, to understand their impact on biochemical processes (Kunsagi-Mate S et al., 2021).

  • Perchlorate detection via an invertebrate biosensor.: Discusses the development of an invertebrate biosensor for the detection of perchlorates, offering a novel approach to monitoring environmental and potentially harmful residues of potassium perchlorate (Sturzenbaum S et al., 2021).

  • Towards understanding the effects of heat and humidity on ageing of a NASA standard pyrotechnic igniter.: Provides insights into the degradation processes of pyrotechnic igniters under varying environmental conditions, useful for enhancing the safety and effectiveness of devices containing potassium perchlorate (Yoh JJ et al., 2019).

  • Interpreting the near infrared region of explosives.: Analyzes the near-infrared spectral characteristics of various explosives, including studies on potassium perchlorate, to improve detection and safety protocols in analytical chemistry applications (Garcia-Ruiz C et al., 2018).

Analysis Note

Assay (alkalimetric): 99.5 - 100.5 %
Insoluble matter: ≤ 0.005 %
pH-value (1 %; water): 5.0 - 7.0
Chloride (Cl): ≤ 0.003 %
Sulfate (SO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service