Skip to Content
Merck

1.04820

SAFC

Potassium acetate

EMPROVE® ESSENTIAL, Ph. Eur., BP, JPE, E 261

Pharma Manufacturing

Synonym(s):

Potassium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3KO2
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12352107
E Number:
E261
EC Index Number:
204-822-2

Agency

BP
JPE
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

quality

E 261

pH

7.5-8.5 (20 °C, 50 g/L in H2O)

mp

292 °C

density

1.57 g/cm3 at 25 °C

application(s)

liquid formulation
ophthalmics
pharma/biopharma processes
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[K+].[O-]C(=O)C

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium acetate extra pure is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service